missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta Galactosidase Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. A11132
Encompass
Modifier l'affichage
Click to view available options
Quantité:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A11132 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A11132 Fournisseur Invitrogen™ Code fournisseur A11132

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

The E. coli gene lacZ encoding beta-galactosidase is an important reporter gene for detecting the expression of recombinant genes in animal cells. In situations where enzymatic detection of beta-galactosidase is not practical, anti-beta-galactosidase antibody has been widely used. Anti-beta-galactosidase antibody can also be used for purification of fusion proteins by immunoprecipitation or immunoadsorption. Furthermore, anti-beta-galactosidase antibody has been shown to be the preferred analytical tool for light microscopic evaluation of lacZ gene transfer, and the beta-galactosidase/anti-beta-galactosidase complex has been used as a model to assess the feasibility of a high-pressure immunodesorption process. This antibody, raised in rabbit against E. coli beta-galactosidase, demonstrates high specificity for the enzyme and is suitable for immunological methods that require high titer and specificity such as immunoblotting (western or dot blot), ELISA, immunoprecipitation, and immunohistochemistry. The appropriate dilution of antibody for a particular application should be determined empirically. We suggest trying dilutions of about 1:5,000 to 1:50,000 for your initial experiments.

Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.

Spécifications

Antigène beta Galactosidase
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugué Unconjugated
Formule PBS with 5mM sodium azide; pH 7.2
Alias du gène acid beta-galactosidase; AW125515; betagal; Beta-Gal; Beta-galactosidase; bgal; Bge; Bgl; Bgl-e; Bgl-s; Bgl-t; Bgs; Bgt; C130097A14Rik; EBP; Elastin receptor 1; elastin receptor 1, 67kDa; ELNR1; galactosidase beta 1; galactosidase, beta 1; GLB1; Glb-1; lactase; lysosmal hydrolase; lysosomal beta-galactosidase; lysosomal hydrolase; MPS4B; ßgal; ß-gal; ß-galactosidase
Espèces hôtes Rabbit
Immunogène beta-Galactosidase.
Méthode de purification IgG fraction
Quantité 500 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Bacteria
Contenu et stockage 4°C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats