missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Thioglucose sodium salt, 95%

Catalog No. AAJ66838MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66838MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66838MD Supplier Thermo Scientific Chemicals Supplier No. J66838MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10593-29-0 | C6H11NaO5S | 218.199 g/mol

beta-D-Thioglucose sodium salt, is used in research and to treat rheumatoid arthritis. The biological mechanism is unknown. 1-Thio-β is often used as a carrier molecule for cell uptake of markers, polymers, and nanoparticles, and potentially useful synthetic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-D-Thioglucose sodium salt, is used in research and to treat rheumatoid arthritis. The biological mechanism is unknown. 1-Thio-β is often used as a carrier molecule for cell uptake of markers, polymers, and nanoparticles, and potentially useful synthetic reagent.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 10593-29-0
Molecular Formula C6H11NaO5S
Molecular Weight (g/mol) 218.199
MDL Number MFCD00061630
InChI Key VKPBZIVFRYLHPT-WNFIKIDCSA-M
Synonym sodium glucopyranosyl sulphide, glucothiose, sodium 2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-thiolate, 1-thio-beta-d-glucose sodium salt, beta-d-thioglucose, 1-, a-d-glucosylthiose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-sodiosulfanyl oxane-3,4,5-triol, sodium 2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl sulfanide, 1-thio-, a-d-glucose sodium salt, 1-beta-d-glucosylthiose
PubChem CID 11413308
IUPAC Name sodium;(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxane-2-thiolate
SMILES C(C1C(C(C(C(O1)[S-])O)O)O)O.[Na+]

Specifications

Color White
Quantity 250 mg
Solubility Information Soluble in water
Formula Weight 218.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material beta-D-Thioglucose sodium salt
EINECSNumber 234-200-6
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only