missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-D-Glucose (contains alpha-D-Glucose) 85.0+%, TCI America™
Specifications
Specifications
| Melting Point | 155°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 180.16 |
| Percent Purity | ≥85.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | beta-D-Glucose (contains alpha-D-Glucose) |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Refrigerator |