missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclodextrin 99.0+%, TCI America™

Catalog No. C077725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C077725G 25 g
C0777100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C077725G Supplier TCI America Supplier No. C077725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7585-39-9
Molecular Formula C42H70O35
Molecular Weight (g/mol) 1134.99
MDL Number MFCD00078139
InChI Key WHGYBXFWUBPSRW-UHFFFAOYNA-N
Synonym cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose
PubChem CID 131707246
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Color White
Quantity 25 g
Formula Weight 1134.99
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material beta-Cyclodextrin
EINECSNumber (8)-0101
RTECSNumber GU2293000
TSCA Yes