missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bêta-Citronellol 95,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 220°C |
| Quantity | 25 mL |
| Formula Weight | 156.27 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | beta-Citronellol |
Sécurité et manipulation
| EINECSNumber | (2)-0258 |
| RTECSNumber | RH3400000 |
| TSCA | Yes |