missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 198°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 89.09 |
| Percent Purity | ≥99.0% (T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | beta-Alanine |
Safety and Handling
| EINECSNumber | (9)-1554 |
| TSCA | Yes |