missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine 99.0+%, TCI America™

Catalog No. A018025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A018025G 25 g
A0180500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A018025G Supplier TCI America Supplier No. A018025G

Chemical Identifiers

CAS 107-95-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008200
InChI Key UCMIRNVEIXFBKS-UHFFFAOYSA-N
Synonym beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
PubChem CID 239
ChEBI CHEBI:16958
IUPAC Name 3-aminopropanoic acid
SMILES NCCC(O)=O

Specifications

Melting Point 198°C
Color White
Quantity 25 g
Formula Weight 89.09
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanine
EINECSNumber (9)-1554
TSCA Yes