missing translation for 'onlineSavingsMsg'
Learn More

beta,3-Dinitrostyrene, 98%

Catalog No. AAL1230406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1230406 5 g
AAL1230403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1230406 Supplier Thermo Scientific Chemicals Supplier No. L1230406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 882-26-8 | C8H6N2O4 | 194.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 882-26-8
Molecular Formula C8H6N2O4
Molecular Weight (g/mol) 194.146
MDL Number MFCD00052063
InChI Key YOEGXQQUPVDQEE-SNAWJCMRSA-N
Synonym beta,3-dinitrostyrene, 1-nitro-3-2-nitrovinyl benzene, m,.beta.-dinitrostyrene, trans-m,beta-dinitrostyrene, m-2-nitrovinyl nitrobenzene, 3-nitro-b-nitrostyrene, styrene, m,beta-dinitro-, e, benzene, 1-nitro-3-2-nitroethenyl, 1-nitro-2-m-nitrophenyl ethylene, 1-nitro-3-e-2-nitroethenyl benzene
PubChem CID 5355017
IUPAC Name 1-nitro-3-[(E)-2-nitroethenyl]benzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C=C[N+](=O)[O-]

Specifications

Melting Point 125°C to 127°C
Quantity 5 g
Beilstein 2051290
Formula Weight 194.15
Percent Purity 98%
Chemical Name or Material beta,3-Dinitrostyrene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only