Learn More
Abcam Inc Bestatin hydrochloride, aminopeptidase inhibitor, 5MG
Numéro de catalogue. 502706253
Encompass
MW 344.8 Da, Purity >98%. Potent and specific aminopeptidase inhibitor. Antibiotic. Inhibits 12 aminopeptidases including aminopeptidase B, (Ki = 1 M), leucyl aminopeptidase (Ki = 1 nM) and cytosol nonspecific dipeptidase (Ki = 4 nM). Active in vivo and in vitro..
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502706253 Fournisseur Abcam Inc Code fournisseur AB1424055MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 344.8 Da, Purity >98%. Potent and specific aminopeptidase inhibitor. Antibiotic. Inhibits 12 aminopeptidases including aminopeptidase B, (Ki = 1 M), leucyl aminopeptidase (Ki = 1 nM) and cytosol nonspecific dipeptidase (Ki = 4 nM). Active in vivo and in vitro..
The product is subject to the following: Abcam Restricted Use Statement