missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BES sodium salt, 99%

Numéro de catalogue. AC439035000
Modifier l'affichage
Click to view available options
500g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
AC439035000 500g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC439035000 Fournisseur Thermo Scientific™ Code fournisseur 439035000

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer

Spécifications

Nom ou substance chimique BES sodium salt
CAS 66992-27-6
CAS % minimum 98.5
CAS % maximal 100.0
Quantité 500g
Fourchette de pourcentages d’essai 99%
Formule moléculaire C6H14NO5S·Na
Synonyme bes sodium salt, sodium 2-bis 2-hydroxyethyl amino ethanesulfonate, n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid sodium salt, sodium 2-bis 2-hydroxyethyl amino ethane-1-sulfonate, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino-, monosodium salt, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, sodium salt, bes na, bes sodium salt buffer, potassium c6h14no5s
Clé InChI CFQLQLSIZOWFNV-UHFFFAOYSA-M
SOURIRES C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]
Nom de l’IUPAC sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
Poids moléculaire (g/mol) 235.24
PubChem CID 23671640
Poids de formule 235.24
Pourcentage de pureté 99%
Spectre infrarouge Authentic
Afficher plus de résultats Afficher moins de résultats
Hazard Statement GHS H Statement:
Precautionary Statement GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only