missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BES, 99+%, for biochemistry

Catalog No. p-65622
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172580250 25 g Glass bottle
AC172581000 100 g Glass bottle
AC172585000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC172580250 Supplier Thermo Scientific Chemicals Supplier No. 172580250
Only null left

CAS: 10191-18-1 | C6H15NO5S | 213.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N, N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid
PubChem CID 73243
ChEBI CHEBI:39045
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Absorbance 0.095 max. (1 M in H2O) at 260nm
Melting Point 152.0°C to 154.0°C
CAS Min % 99.0
Color Beige to Brown
CAS Max % 100.0
Physical Form Powder
Quantity 25 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Loss on Drying 0.5 % max. (100°C, 2 hrs)
Linear Formula (HOCH2CH2)2NCH2CH2SO3H
Packaging Glass bottle
Solubility Information Solubility in water: soluble.
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
Formula Weight 213.25
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material BES
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-465-5
RTECSNumber KI7363500
TSCA TSCA

RUO – Research Use Only