accessibility menu, dialog, popup

UserName

HONOHSOHOO

CAS RN 10191-18-1

IUPAC Name: 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
Synonyms: N,N-bis(2-hydroxyethyl)taurine bes (buffering agent) BES
Molecular Weight (g/mol): 213.25
Molecular Formula: C6H15NO5S
InChi Key: AJTVSSFTXWNIRG-UHFFFAOYSA-N
SMILES: OCCN(CCO)CCS(O)(=O)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)