missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Organometallic Standards, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC ORGBE82Z

Catalog No. ORGBE82Z


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Be1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Beryllium Oil Standard
7440-41-7 , 64742-65-0
0.1,99.9
Solid
MFCD00134032
Certified Reference Material
[Be]
beryllium
9.01
5460467
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,99.9
Be
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
Base Oil 75
Petrochemical Testing
50 g
CHEBI:33783

Chemical Identifiers

7440-41-7
9.01
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
CHEBI:33783
[Be]
Be
MFCD00134032
5460467
beryllium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only