missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium Tribromide 97.0+%, TCI America™

Catalog No. T138225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T138225G 25 g
T13825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T138225G Supplier TCI America Supplier No. T138225G

Chemical Identifiers

CAS 111865-47-5
Molecular Formula C10H17Br3N+
Molecular Weight (g/mol) 390.965
MDL Number MFCD00134423
InChI Key GSDDWZVWOXMLJZ-UHFFFAOYSA-N
Synonym benzyltrimethylammonium tribromide
PubChem CID 86222689
IUPAC Name benzyl(trimethyl)azanium;molecular bromine;hydrobromide
SMILES C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr

Specifications

Melting Point 102°C
Quantity 25 g
Formula Weight 389.96
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltrimethylammonium Tribromide [Brominating Reagent]
EINECSNumber (3)-2694
TSCA Yes