missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium hydroxide, 40% w/w in methanol

Catalog No. AAA14927AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14927AP 500 mL
AAA14927AE 100 mL
AAA149270F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA14927AP Supplier Thermo Scientific Chemicals Supplier No. A14927AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-85-6 | C10H17NO | 167.252 g/mol

Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.

Solubility
Miscible with water, hydrocarbons, aromatic hydrocarbons and halogenated solvents.

Notes
Air Sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 100-85-6
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00008281
InChI Key NDKBVBUGCNGSJJ-UHFFFAOYSA-M
Synonym benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
PubChem CID 66854
IUPAC Name benzyl(trimethyl)azanium;hydroxide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[OH-]

Specifications

Density 0.92
Boiling Point 65°C
Flash Point 15°C (59°F)
Odor Ammonia-like
Refractive Index 1.427
Linear Formula C6H5CH2N(CH3)3OH
Quantity 500 mL
UN Number UN2924
Beilstein 3917256
Sensitivity Air Sensitive
Solubility Information Miscible with water,hydrocarbons,aromatic hydrocarbons and halogenated solvents.
Formula Weight 167.25
Concentration or Composition (by Analyte or Components) 40% w/w in methanol
Chemical Name or Material Benzyltrimethylammonium hydroxide
Show More Show Less
Hazard Category H225-H301+H311+H331-H314-H335-H370-H372
Hazard Statement GHS H Statement
H225-H311-H331-H370-H314
Highly flammable liquid and vapour.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 202-895-5
RTECSNumber BO8575000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only