missing translation for 'onlineSavingsMsg'
Learn More

Benzyltri-n-propylammonium chloride, 96%

Catalog No. AAL0067509
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0067509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0067509 Supplier Thermo Scientific Chemicals Supplier No. L0067509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5197-87-5 | C16H28ClN | 269.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5197-87-5
Molecular Formula C16H28ClN
Molecular Weight (g/mol) 269.86
MDL Number MFCD00051869
InChI Key YTRIOKYQEVFKGU-UHFFFAOYSA-M
Synonym benzyltripropylammonium chloride, n-benzyl-n,n-dipropylpropan-1-aminium chloride, benzyl tripropyl ammonium chloride, benzyltri-n-propylammonium chloride, benzyltripropylazanium chloride, benzyl tripropyl azanium chloride, acmc-20alov, c16h28n.cl, benzyl-tri-n-propylammonium chloride
PubChem CID 6097067
IUPAC Name benzyltripropylazanium chloride
SMILES [Cl-].CCC[N+](CCC)(CCC)CC1=CC=CC=C1

Specifications

Melting Point ∽190°C (decomposition)
Linear Formula C6H5CH2N(CH2CH2CH3)3Cl
Quantity 10 g
Beilstein 4280671
Sensitivity Hygroscopic
Formula Weight 269.86
Percent Purity 96%
Chemical Name or Material Benzyltri-n-propylammonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only