missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetic acid, 95%, Thermo Scientific™

Catalog No. p-4813341
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC427080010 1g
AC427080050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427080010 Supplier Thermo Scientific Chemicals Supplier No. 427080010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 30379-55-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00274211
InChI Key GRZHHTYDZVRPIC-UHFFFAOYSA-N
Synonym benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy
PubChem CID 290301
IUPAC Name 2-phenylmethoxyacetic acid
SMILES OC(=O)COCC1=CC=CC=C1

Specifications

Density 1.1620g/mL
Boiling Point 137°C to 139°C (0.6mmHg)
Flash Point 113°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2COOH
Quantity 1g
Specific Gravity 1.162
Formula Weight 166.18
Percent Purity 95%
Chemical Name or Material Benzyloxyacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only