missing translation for 'onlineSavingsMsg'
Learn More

Benzylmalonic Acid 98.0+%, TCI America™

Numéro de catalogue. B04225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B04225G 5 g
B042225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B04225G Fournisseur TCI America Code fournisseur B04225G
Il en reste null

Identifiants chimiques

CAS 616-75-1
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004261
InChI Key JAEJSNFTJMYIEF-UHFFFAOYSA-N
PubChem CID 12031
IUPAC Name 2-benzylpropanedioic acid
SMILES OC(=O)C(CC1=CC=CC=C1)C(O)=O

Spécifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylmalonic Acid
RTECSNumber OO0525000
TSCA No