missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylphenylammonium Chloride 98.0+%, TCI America™

Supplier:  TCI America B041625G

Catalog No. B041625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Benzyldimethylphenylammonium Chloride
White
MFCD00031566
benzyldimethylphenylammonium chloride, leukotrope, n-benzyl-n,n-dimethylanilinium chloride, leucotrope o, dimethylbenzylanilinium chloride, benzenemethanaminium, n,n-dimethyl-n-phenyl-, chloride, benzyldimethylanilinium chloride, dimethylphenylbenzylammonium chloride, phenyldimethylbenzylammonium chloride, n,n-dimethyl-n-phenylbenzenemethanaminium chloride
[Cl-].C[N+](C)(CC1=CC=CC=C1)C1=CC=CC=C1
247.77
247.77
Crystalline Powder
3204-68-0
C15H18ClN
25 g
QLRKASHXFNIPLZ-UHFFFAOYSA-M
N-benzyl-N,N-dimethylanilinium chloride
76668
≥98.0% (HPLC,T)

Chemical Identifiers

3204-68-0
247.77
QLRKASHXFNIPLZ-UHFFFAOYSA-M
76668
[Cl-].C[N+](C)(CC1=CC=CC=C1)C1=CC=CC=C1
C15H18ClN
MFCD00031566
benzyldimethylphenylammonium chloride, leukotrope, n-benzyl-n,n-dimethylanilinium chloride, leucotrope o, dimethylbenzylanilinium chloride, benzenemethanaminium, n,n-dimethyl-n-phenyl-, chloride, benzyldimethylanilinium chloride, dimethylphenylbenzylammonium chloride, phenyldimethylbenzylammonium chloride, n,n-dimethyl-n-phenylbenzenemethanaminium chloride
N-benzyl-N,N-dimethylanilinium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions