missing translation for 'onlineSavingsMsg'
Learn More

Benzylamine 99.0+%, TCI America™

Catalog No. B0406500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0406500ML 500 mL
B040625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0406500ML Supplier TCI America Supplier No. B0406500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-46-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00008106
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN

Specifications

Color Yellow
Boiling Point 184°C
Quantity 500 mL
UN Number 2734
Formula Weight 107.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzylamine
EINECSNumber (3)-0367
RTECSNumber DP1488500
TSCA Yes