missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylamine Hydroiodide (Low water content) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 178°C |
| Quantity | 5 g |
| Formula Weight | 235.07 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzylamine Hydroiodide (Low water content) |
Safety and Handling
| TSCA | No |