missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylamine Hydrobromide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 204°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 188.07 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzylamine Hydrobromide |
Safety and Handling
| TSCA | No |