missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Formate 95.0+%, TCI America™
$55.92 - $354.17
Chemical Identifiers
| CAS | 104-57-4 |
|---|---|
| Molecular Formula | C8H8O2 |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00014128 |
| InChI Key | UYWQUFXKFGHYNT-UHFFFAOYSA-N |
| Synonym | formic acid, phenylmethyl ester, benzyl methanoate, phenylmethyl formate, benzyl alcohol, formate, formic acid benzyl ester, benzyl formiat, formic acid, benzyl ester, benzylester kyseliny mravenci, ameisensaeurebenzylester, carbobenzoxy group |
| PubChem CID | 7708 |
| ChEBI | CHEBI:3057 |
| IUPAC Name | benzyl formate |
| SMILES | C1=CC=C(C=C1)COC=O |
Chemical Identifiers
| 104-57-4 | |
| 136.15 | |
| UYWQUFXKFGHYNT-UHFFFAOYSA-N | |
| 7708 | |
| benzyl formate |
| C8H8O2 | |
| MFCD00014128 | |
| formic acid, phenylmethyl ester, benzyl methanoate, phenylmethyl formate, benzyl alcohol, formate, formic acid benzyl ester, benzyl formiat, formic acid, benzyl ester, benzylester kyseliny mravenci, ameisensaeurebenzylester, carbobenzoxy group | |
| CHEBI:3057 | |
| C1=CC=C(C=C1)COC=O |
Specifications
| 104-57-4 | |
| 201°C | |
| MFCD00014128 | |
| formic acid, phenylmethyl ester, benzyl methanoate, phenylmethyl formate, benzyl alcohol, formate, formic acid benzyl ester, benzyl formiat, formic acid, benzyl ester, benzylester kyseliny mravenci, ameisensaeurebenzylester, carbobenzoxy group | |
| C1=CC=C(C=C1)COC=O | |
| 136.15 | |
| CHEBI:3057 | |
| ≥95.0% (GC) | |
| Benzyl Formate |
| Colorless | |
| C8H8O2 | |
| 25 g | |
| UYWQUFXKFGHYNT-UHFFFAOYSA-N | |
| benzyl formate | |
| 7708 | |
| 136.15 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-1020
RTECSNumber : LQ5400000
TSCA : Yes