missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Formate 95.0+%, TCI America™

Supplier:  TCI America F0050500G

 View more versions of this product

Catalog No. F0050500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Benzyl Formate
Colorless
C8H8O2
500 g
UYWQUFXKFGHYNT-UHFFFAOYSA-N
benzyl formate
7708
136.15
Liquid
104-57-4
201°C
MFCD00014128
formic acid, phenylmethyl ester, benzyl methanoate, phenylmethyl formate, benzyl alcohol, formate, formic acid benzyl ester, benzyl formiat, formic acid, benzyl ester, benzylester kyseliny mravenci, ameisensaeurebenzylester, carbobenzoxy group
C1=CC=C(C=C1)COC=O
136.15
CHEBI:3057
≥95.0% (GC)

Chemical Identifiers

104-57-4
136.15
UYWQUFXKFGHYNT-UHFFFAOYSA-N
7708
benzyl formate
C8H8O2
MFCD00014128
formic acid, phenylmethyl ester, benzyl methanoate, phenylmethyl formate, benzyl alcohol, formate, formic acid benzyl ester, benzyl formiat, formic acid, benzyl ester, benzylester kyseliny mravenci, ameisensaeurebenzylester, carbobenzoxy group
CHEBI:3057
C1=CC=C(C=C1)COC=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1020

RTECSNumber : LQ5400000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions