accessibility menu, dialog, popup

UserName

OO

CAS RN 104-57-4

IUPAC Name: benzyl formate
Synonyms: formic acid benzyl ester ameisensaeurebenzylester benzylester kyseliny mravenci benzyl formiat
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: UYWQUFXKFGHYNT-UHFFFAOYSA-N
SMILES: O=COCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results