missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Formate 95.0+%, TCI America™
Supplier: TCI America F005025G
Specifications
| Benzyl Formate | |
| Colorless | |
| C8H8O2 | |
| 25 g | |
| UYWQUFXKFGHYNT-UHFFFAOYSA-N | |
| benzyl formate | |
| 7708 | |
| 136.15 | |
| Liquid |
| 104-57-4 | |
| 201°C | |
| MFCD00014128 | |
| formic acid, phenylmethyl ester, benzyl methanoate, phenylmethyl formate, benzyl alcohol, formate, formic acid benzyl ester, benzyl formiat, formic acid, benzyl ester, benzylester kyseliny mravenci, ameisensaeurebenzylester, carbobenzoxy group | |
| C1=CC=C(C=C1)COC=O | |
| 136.15 | |
| CHEBI:3057 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 104-57-4 | |
| 136.15 | |
| UYWQUFXKFGHYNT-UHFFFAOYSA-N | |
| 7708 | |
| benzyl formate |
| C8H8O2 | |
| MFCD00014128 | |
| formic acid, phenylmethyl ester, benzyl methanoate, phenylmethyl formate, benzyl alcohol, formate, formic acid benzyl ester, benzyl formiat, formic acid, benzyl ester, benzylester kyseliny mravenci, ameisensaeurebenzylester, carbobenzoxy group | |
| CHEBI:3057 | |
| C1=CC=C(C=C1)COC=O |
Safety and Handling
EINECSNumber : (3)-1020
RTECSNumber : LQ5400000
TSCA : Yes