missing translation for 'onlineSavingsMsg'
Learn More

Benzyl(ethyl)dimethylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. B54275G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B54275G Supplier TCI America Supplier No. B54275G
Only null left

Chemical Identifiers

CAS 1186103-43-4
Molecular Formula C13H18F6N2O4S2
Molecular Weight (g/mol) 444.407
InChI Key IGABCHQKFATAOI-UHFFFAOYSA-N
PubChem CID 87176851
IUPAC Name benzyl-ethyl-dimethylazanium;bis(trifluoromethylsulfonyl)azanide
SMILES CC[N+](C)(C)CC1=CC=CC=C1.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 444.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material Benzyl(ethyl)dimethylammonium Bis(trifluoromethanesulfonyl)imide