missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Chloroacetate 97.0+%, TCI America™

Catalog No. C126525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C126525G 25 g
C1265250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C126525G Supplier TCI America Supplier No. C126525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140-18-1
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00157062
InChI Key SOGXBRHOWDEKQB-UHFFFAOYSA-N
Synonym benzyl chloroacetate, benzyl monochloracetate, acetic acid, chloro-, phenylmethyl ester, chloroacetic acid, benzyl ester, monochloroacetate, chloroacetic acid benzyl ester, benzyl-alpha-chloroacetate, phenylmethyl chloroacetate, acetic acid, chloro-, benzyl ester, phenylmethyl 2-chloroacetate
PubChem CID 8786
IUPAC Name benzyl 2-chloroacetate
SMILES C1=CC=C(C=C1)COC(=O)CCl

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 g
Formula Weight 184.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Chloroacetate
RTECSNumber AF8750000
TSCA Yes