Learn More
Benzyl chloride, 99%, stab.
Description
Benzyl Chloride is used as an intermediate for manufacturing of benzyl alcohol, benzoic acid and benzyl cyanide. It is also used to prepare benzyl esters, which are used as plasticizer, flavorants and perfumes. It acts as an intermediate in the preparation of phenylacetic acid. In organic synthesis, it is used as an alkylating agent and utilized for the introduction of the benzyl protecting group for alcohols. Further, it is involved in the preparation of quaternary ammonium salts by reaction with tertiary amines. In addition to this, it is used to prepare Grignard reagent by reacting with metallic magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzyl Chloride is used as an intermediate for manufacturing of benzyl alcohol, benzoic acid and benzyl cyanide. It is also used to prepare benzyl esters, which are used as plasticizer, flavorants and perfumes. It acts as an intermediate in the preparation of phenylacetic acid. In organic synthesis, it is used as an alkylating agent and utilized for the introduction of the benzyl protecting group for alcohols. Further, it is involved in the preparation of quaternary ammonium salts by reaction with tertiary amines. In addition to this, it is used to prepare Grignard reagent by reacting with metallic magnesium.
Solubility
Miscible with alcohol, ether. Insoluble in waterMiscible with alcohol, ether and chloroform.
Notes
Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with common metals, lead,oxidizing agents, iron, iron salts, brass and aluminum
Spécifications
Spécifications
| Melting Point | -43°C |
| Density | 1.099 |
| Boiling Point | 178°C to 180°C |
| Flash Point | 73°C (163°F) |
| Assay Percent Range | 99%, Stabilized |
| Refractive Index | 1.538 |
| Quantity | 10,000 g |
| UN Number | UN1738 |
| Beilstein | 471308 |
| Merck Index | 14,1129 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H227-H290-H302-H315-H317-H318-H331-H335-H336-H340-H350-H373 |
| Hazard Statement | GHS H Statement H301-H330-H350-H373-H314-H318-H227-H335 Toxic if swallowed. Fatal if inhaled. May cause cancer. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P234-P235-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P333+P313-P362-P370+P378q-P390-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZYL CHLORIDE |
| EINECSNumber | 202-853-6 |
| RTECSNumber | XS8925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only