missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol, ACS Reagent, Reagents
Benzyl Alcohol, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C215208055E
Specifications
| Benzyl Alcohol, ACS Reagent | |
| 205°C | |
| Colorless | |
| 205°C | |
| Steel Drum | |
| 55 gal. | |
| Phenylcarbinol, Phenylmethanol, Benzenemethanol | |
| OCC1=CC=CC=C1 | |
| 108.14 | |
| Liquid |
| 100-51-6 | |
| -15.4°C | |
| Neutral | |
| -15.4°C | |
| C7H8O | |
| 1.044 | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| phenylmethanol | |
| ACS |
Chemical Identifiers
| 100-51-6 | |
| 108.14 | |
| Phenylcarbinol, Phenylmethanol, Benzenemethanol | |
| OCC1=CC=CC=C1 |
| C7H8O | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| phenylmethanol |