missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, ACS Reagent, Reagents

Catalog No. p-200021137
Change view
Click to view available options
Quantity:
4 L
15 gal.
55 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337469 15 gal.
01337468 4 L
01337467 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337469 Supplier Labchem Inc Supplier No. C215208015E
Only null left

Benzyl Alcohol, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym Phenylcarbinol, Phenylmethanol, Benzenemethanol
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Boiling Range 205°C
Melting Point -15.4°C
Color Colorless
pH Neutral
Boiling Point 205°C
Melting Range -15.4°C
Packaging Steel Drum
Quantity 15 gal.
Specific Gravity 1.044
Grade ACS
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol, ACS Reagent
Show More Show Less