missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 5-Bromoamyl Ether 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 257.17 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Benzyl 5-Bromoamyl Ether |
Safety and Handling
| TSCA | No |