missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (3-Fluoro-4-morpholinophenyl)carbamate 98.0+%, TCI America™

Catalog No. B48105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48105G 5 g
B481025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48105G Supplier TCI America Supplier No. B48105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 168828-81-7
Molecular Formula C18H19FN2O3
Molecular Weight (g/mol) 330.359
MDL Number MFCD06658240
InChI Key XKGUZGHMWUIYDR-UHFFFAOYSA-N
Synonym (3-Fluoro-4-morpholinophenyl)carbamic Acid Benzyl Ester
PubChem CID 5306150
IUPAC Name benzyl N-(3-fluoro-4-morpholin-4-ylphenyl)carbamate
SMILES C1COCCN1C2=C(C=C(C=C2)NC(=O)OCC3=CC=CC=C3)F

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 5 g
Formula Weight 330.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl (3-Fluoro-4-morpholinophenyl)carbamate