missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 3-Bromopropyl Ether 95.0+%, TCI America™

Catalog No. B28185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28185G 5 g
B281825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28185G Supplier TCI America Supplier No. B28185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54314-84-0
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.12
MDL Number MFCD00134570
InChI Key PSUXTZLDBVEZTD-UHFFFAOYSA-N
Synonym benzyl 3-bromopropyl ether, 3-bromopropoxy methyl benzene, 3-bromopropylbenzyl ether, 3-benzyloxy-1-bromopropane, 3-benzyloxy propyl bromide, benzene, 3-bromopropoxy methyl, 1-3-bromopropoxy methyl benzene, benzyl3-bromopropylether
PubChem CID 2776064
IUPAC Name [(3-bromopropoxy)methyl]benzene
SMILES BrCCCOCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 141°C
Quantity 5 g
Formula Weight 229.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl 3-Bromopropyl Ether