missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 3-bromopropyl ether, 98%

Catalog No. AC399130100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399130100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399130100 Supplier Thermo Scientific Chemicals Supplier No. 399130100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54314-84-0 | C10H13BrO | 229.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54314-84-0
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.12
MDL Number MFCD00134570
InChI Key PSUXTZLDBVEZTD-UHFFFAOYSA-N
Synonym benzyl 3-bromopropyl ether, 3-bromopropoxy methyl benzene, 3-bromopropylbenzyl ether, 3-benzyloxy-1-bromopropane, 3-benzyloxy propyl bromide, benzene, 3-bromopropoxy methyl, 1-3-bromopropoxy methyl benzene, benzyl3-bromopropylether
PubChem CID 2776064
IUPAC Name 3-bromopropoxymethylbenzene
SMILES BrCCCOCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.2980g/mL
Boiling Point 130°C to 132°C (8.0 mmHg)
Flash Point 113°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Specific Gravity 1.298
Formula Weight 229.11
Percent Purity 98%
Chemical Name or Material Benzyl 3-bromopropyl ether
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only