missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-bromoethyl ether, 97%, Thermo Scientific™

Catalog No. p-3586398
Change view
Click to view available options
Quantity:
5g
25g
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC388870050 5g
AC388870250 25g
AC388871000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC388870050 Supplier Thermo Scientific Chemicals Supplier No. 388870050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1462-37-9
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD01321307
InChI Key FWOHDAGPWDEWIB-UHFFFAOYSA-N
Synonym benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene
PubChem CID 73833
IUPAC Name 2-bromoethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.3600g/mL
Boiling Point 254.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2CH2Br
Quantity 5g
Specific Gravity 1.36
Formula Weight 215.09
Percent Purity 97%
Chemical Name or Material Benzyl 2-bromoethyl ether
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only