missing translation for 'onlineSavingsMsg'
Learn More

Benzoyleneurea 98.0+%, TCI America™

Catalog No. B33815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33815G 5 g
B338125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33815G Supplier TCI America Supplier No. B33815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86-96-4
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00006699
InChI Key SDQJTWBNWQABLE-UHFFFAOYSA-N
Synonym benzoyleneurea, quinazoline-2,4 1h,3h-dione, 2,4 1h,3h-quinazolinedione, quinazoline-2,4-diol, 2,4-dihydroxyquinazoline, quinazolinedione, benzouracil, quinazoline-2,4-dione, urea, benzoylene, 1,2,3,4-tetrahydroquinazoline-2,4-dione
PubChem CID 64048
IUPAC Name 1H-quinazoline-2,4-dione
SMILES C1=CC=C2C(=C1)C(=O)NC(=O)N2

Specifications

Melting Point 354°C
Color White-Yellow
Quantity 5 g
Formula Weight 162.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyleneurea
RTECSNumber VA1390000
TSCA No