missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Chloride 98.0+%, TCI America™

Catalog No. B0105500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0105500ML 500 mL
B010525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0105500ML Supplier TCI America Supplier No. B0105500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-88-4
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.566
MDL Number MFCD00000653
InChI Key PASDCCFISLVPSO-UHFFFAOYSA-N
Synonym benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PubChem CID 7412
ChEBI CHEBI:82275
IUPAC Name benzoyl chloride
SMILES C1=CC=C(C=C1)C(=O)Cl

Specifications

Color Colorless
Boiling Point 196°C
Quantity 500 mL
UN Number 1736
Formula Weight 140.57
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Benzoyl Chloride
EINECSNumber (3)-1387
RTECSNumber DM6600000
TSCA Yes