missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone hydrazone, 98+%, Thermo Scientific™

$101.73 - $243.49

Chemical Identifiers

CAS 5350-57-2
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.25
MDL Number MFCD00007624
InChI Key QYCSNMDOZNUZIT-UHFFFAOYSA-N
Synonym benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
PubChem CID 79304
IUPAC Name benzhydrylidenehydrazine
SMILES C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC105570050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC105571000
View Documents Promotion Details
N/A 100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC105575000
View Documents Promotion Details
N/A 500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5350-57-2
196.25
QYCSNMDOZNUZIT-UHFFFAOYSA-N
79304
C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2
C13H12N2
MFCD00007624
benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
benzhydrylidenehydrazine

Specifications

5350-57-2
100.0
Authentic
C13H12N2
MFCD00007624
13,242
QYCSNMDOZNUZIT-UHFFFAOYSA-N
benzhydrylidenehydrazine
79304
98+%
98.0
225°C to 230°C (55.0mmHg)
98+%
(C6H5)2C=NNH2
07, 417
benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2
196.25
196.25
Benzophenone hydrazone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-321-8

SDS
Documents

Documents

RUO – Research Use Only