missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone hydrazone, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 105570050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC105570050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5350-57-2
196.25
QYCSNMDOZNUZIT-UHFFFAOYSA-N
79304
C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2
C13H12N2
MFCD00007624
benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
benzhydrylidenehydrazine

Specifications

Benzophenone hydrazone
98.0
225°C to 230°C (55.0mmHg)
98+%
C13H12N2
5g
07, 417
benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2
196.25
196.25
5350-57-2
100.0
Authentic
Glass bottle
(C6H5)2C=NNH2
MFCD00007624
13,242
QYCSNMDOZNUZIT-UHFFFAOYSA-N
benzhydrylidenehydrazine
79304
98+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-321-8

SDS
Documents

Documents

Promotions

Promotions