accessibility menu, dialog, popup

UserName

NH2N

CAS RN 5350-57-2

IUPAC Name: (diphenylmethylidene)hydrazine
Synonyms: (diphenylmethylidene)hydrazine
Molecular Weight (g/mol): 196.25
Molecular Formula: C13H12N2
InChi Key: QYCSNMDOZNUZIT-UHFFFAOYSA-N
SMILES: NN=C(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)