missing translation for 'onlineSavingsMsg'
Learn More

Benzoin 98.0+%, TCI America™

Catalog No. B0079500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0079500G 500 g
B007925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0079500G Supplier TCI America Supplier No. B0079500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004496
InChI Key ISAOCJYIOMOJEB-UHFFFAOYNA-N
Synonym benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone
PubChem CID 8400
ChEBI CHEBI:17682
IUPAC Name 2-hydroxy-1,2-diphenylethan-1-one
SMILES OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 134°C
Color White-Yellow
Boiling Point 344°C
Quantity 500 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoin
EINECSNumber (9)-1315
RTECSNumber DI1590000
TSCA Yes