missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoguanamine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 228°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 187.21 |
| Percent Purity | ≥99.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzoguanamine |
Safety and Handling
| EINECSNumber | (5)-1028 |
| RTECSNumber | XY7000000 |
| TSCA | Yes |