missing translation for 'onlineSavingsMsg'
Learn More

Benzofurazan, 97%

Catalog No. AAL1122906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1122906 5 g
AAL1122903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1122906 Supplier Thermo Scientific Chemicals Supplier No. L1122906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 273-09-6 | C6H4N2O | 120.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 273-09-6
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.111
MDL Number MFCD00068064
InChI Key AWBOSXFRPFZLOP-UHFFFAOYSA-N
Synonym benzofurazan, benzo c 1,2,5 oxadiazole, benzofurazane, 2,1,3-benzoxadiazol, benzo-2,1,3-oxadiazole, 2-oxa-1,3-diaza-2h-isoindene, 3,5-cyclohexadiene-1,2-dione, dioxime, furazan, pubchem22999, acmc-209gwb
PubChem CID 67501
IUPAC Name 2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C=C1

Specifications

Melting Point 52°C to 55°C
Boiling Point 80°C (20 mmHg)
Flash Point 76°C (169°F)
Quantity 5 g
Beilstein 112261
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material Benzofurazan
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber DM2580000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only