missing translation for 'onlineSavingsMsg'
Learn More

Benzofurazan-5-boronic acid pinacol ester, 97%

Catalog No. AAH2935806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2935806 5 g
AAH2935803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2935806 Supplier Thermo Scientific Chemicals Supplier No. H2935806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1073355-14-2 | C12H15BN2O3 | 246.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073355-14-2
Molecular Formula C12H15BN2O3
Molecular Weight (g/mol) 246.07
MDL Number MFCD09842717
InChI Key MQXANAGMQAYTMV-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo c 1,2,5 oxadiazole, benzofurazan-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzoxadiazole, benzofurazan-5-boronic acid, pinacol ester, benzo c 1,2,5 oxadiazole-5-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzofurazan, benzo c 1,2,5 oxadiazole-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzoxadiazole, benzo c 1,2,5 oxadiazol-5-ylboronic acid pinacol ester, amtb001
PubChem CID 46738004
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzoxadiazole
SMILES CC1(C)OB(OC1(C)C)C1=CC2=NON=C2C=C1

Specifications

Melting Point 87°C to 88°C
Quantity 5 g
Formula Weight 246.07
Percent Purity 97%
Chemical Name or Material Benzofurazan-5-boronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only