missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo(e)pyrene, SPEX CertiPrep™
Supplier: SPEX CertiPrep Group LLC S440
Specifications
| Benzo(e)pyrene | |
| 192-97-2,75-09-2 | |
| 0.1,99.9 | |
| TXVHTIQJNYSSKO-UHFFFAOYSA-N | |
| benzo[e]pyrene | |
| 1 mL | |
| CHEBI:34567 |
| Ampule | |
| 0.1,99.9 | |
| C20H12 | |
| C1=CC=C2C(=C1)C3=CC=CC4=C3C5=C(C=CC=C25)C=C4 | |
| 252.316 | |
| 9128 |
Chemical Identifiers
| 192-97-2 | |
| 252.316 | |
| 9128 | |
| benzo[e]pyrene |
| C20H12 | |
| TXVHTIQJNYSSKO-UHFFFAOYSA-N | |
| CHEBI:34567 | |
| C1=CC=C2C(=C1)C3=CC=CC4=C3C5=C(C=CC=C25)C=C4 |