accessibility menu, dialog, popup

UserName

CAS RN 192-97-2

CAS RN 192-97-2

IUPAC Name: benzo[e]pyrene
Synonyms: benzo(E)pyrene benz(e)pyrene 4,5-benzopyrene 4,5-benzpyrene benzo(l)pyrene 1,2-benzopyrene 1,2-benzo(e)pyrene B(e)P ccris 786 1,2-benzpyrene
Molecular Weight (g/mol): 252.32
Molecular Formula: C20H12
InChi Key: TXVHTIQJNYSSKO-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC=CC3=CC=C4C=CC=C2C4=C13

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results