missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene, 98+%

Catalog No. AAA1232822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1232822 100 g
AAA1232806 5 g
AAA1232814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1232822 Supplier Thermo Scientific Chemicals Supplier No. A1232822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-15-8 | C8H6S | 134.196 g/mol

Benzo[b]thiophene is used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium. It is used in organic as well as pharmaceutical industry as a component in the synthesis of raloxifene, a breast cancer therapeutic, or hydrodesulfurization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene is used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium. It is used in organic as well as pharmaceutical industry as a component in the synthesis of raloxifene, a breast cancer therapeutic, or hydrodesulfurization reactions.

Solubility
Soluble in water (0.13 g/L at 20°C), chloroform and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 95-15-8
Molecular Formula C8H6S
Molecular Weight (g/mol) 134.196
MDL Number MFCD00005864
InChI Key FCEHBMOGCRZNNI-UHFFFAOYSA-N
Synonym benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen
PubChem CID 7221
ChEBI CHEBI:35858
IUPAC Name 1-benzothiophene
SMILES C1=CC=C2C(=C1)C=CS2

Specifications

Melting Point 28°C to 32°C
Density 1.149
Boiling Point 221°C to 222°C
Flash Point 91°C (195°F)
Odor Unpleasant
Quantity 100 g
UN Number UN3077
Beilstein 80580
Merck Index 14,9303
Solubility Information Soluble in water (0.13g/L at 20°C),chloroform and methanol.
Formula Weight 134.2
Percent Purity ≥98%
Physical Form Crystals or Powder or Crystalline Powder or Fused Solid
Chemical Name or Material Benzo[b]thiophene
Show More Show Less
Hazard Category H302
Hazard Statement GHS H StatementH302 Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-395-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only