missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carboxaldehyde, 97%

Catalog No. AAL1934506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1934506 5 g
AAL1934503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1934506 Supplier Thermo Scientific Chemicals Supplier No. L1934506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3541-37-5 | C9H6OS | 162.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3541-37-5
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD01075041
InChI Key NXSVNPSWARVMAY-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde
PubChem CID 736500
IUPAC Name 1-benzothiophene-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2S1

Specifications

Melting Point 30°C to 32°C
Boiling Point 135°C to 136°C (8 mmHg)
Quantity 5 g
Beilstein 114170
Sensitivity Air Sensitive
Formula Weight 162.21
Percent Purity 97%
Chemical Name or Material Benzo[b]thiophene-2-carboxaldehyde
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only