missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-7-sulfonyl chloride, Thermo Scientific™

Chemical Identifiers

CAS 1191030-88-2
Molecular Formula C8H5ClO3S
Molecular Weight (g/mol) 216.635
InChI Key MEOOVUNNRYYGCV-UHFFFAOYSA-N
Synonym benzofuran-7-sulfonyl chloride, 7-benzofuransulfonyl chloride, benzo 2,3-b furan-7-ylchlorosulfone
PubChem CID 49762852
IUPAC Name 1-benzofuran-7-sulfonyl chloride
SMILES C1=CC2=C(C(=C1)S(=O)(=O)Cl)OC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC78203DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC78203DE
View Documents
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1191030-88-2
216.635
benzofuran-7-sulfonyl chloride, 7-benzofuransulfonyl chloride, benzo 2,3-b furan-7-ylchlorosulfone
1-benzofuran-7-sulfonyl chloride
C8H5ClO3S
MEOOVUNNRYYGCV-UHFFFAOYSA-N
49762852
C1=CC2=C(C(=C1)S(=O)(=O)Cl)OC=C2

Specifications

1191030-88-2
benzofuran-7-sulfonyl chloride, 7-benzofuransulfonyl chloride, benzo 2,3-b furan-7-ylchlorosulfone
C1=CC2=C(C(=C1)S(=O)(=O)Cl)OC=C2
216.635
216.64
Benzo[b]furan-7-sulfonyl chloride
C8H5ClO3S
MEOOVUNNRYYGCV-UHFFFAOYSA-N
1-benzofuran-7-sulfonyl chloride
49762852
97%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents